Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pontamine Sky Blue 6BX, MP Biomedicals

Catalog No. icn228230 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN228230 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN228230 Supplier MP Biomedicals, Inc Supplier No. 0522823010

Chemical Identifiers

CAS 2610-10-8
Molecular Formula C45H26N10Na6O21S6
Molecular Weight (g/mol) 1373.05
InChI Key LDCKXVAPQFUIOI-ZFLDVXHKSA-H
PubChem CID 57369623
IUPAC Name hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 10 g
Chemical Name or Material Pontamine Sky Blue 6BX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.