Learn More
Med Vet International Poole Suction Handle, Plastic, J0489PS, 22cm, Each
Catalog No. 5000581835
Encompass_Preferred
Distal 1/3 end is vented with many holes. Fits standard suction hose. Screws apart for easy cleaning. Stainless steel. 8mm diameter. 22cm L. Economy.
Catalog No. 50-005-81835 Supplier Med Vet International Supplier No. JOR489PS
Distal 1/3 end is vented with many holes. Fits standard suction hose. Screws apart for easy cleaning. Stainless steel. 8mm diameter. 22cm L. Economy.