Learn More
Innovative Research Inc Pooled Human AB Serum Plasma Derived 100ml
Catalog No. 502036404
Encompass_Preferred
Pooled Human AB Serum from Innovative Research is collected from male donors with blood type AB. The plasma (collected via apheresis) is processed into serum at FDA-licensed facilities located in the United States. Our Pooled Human AB Serum is filtered and cell culture tested for mycoplasma, endotoxin, and sterility. This product is suitable for use in cell culture applications.
Catalog No. 50-203-6404 Supplier Innovative Research Inc Supplier No. ISERAB100ML
Pooled Human AB Serum from Innovative Research is collected from male donors with blood type AB. The plasma (collected via apheresis) is processed into serum at FDA-licensed facilities located in the United States. Our Pooled Human AB Serum is filtered and cell culture tested for mycoplasma, endotoxin, and sterility. This product is suitable for use in cell culture applications.