Learn More
Innovative Research Inc Pooled Human Complement Plasma, 50ml, K2 EDTA
Catalog No. NC3588015
Encompass_Preferred
Pooled Human Complement Plasma from Innovative Research is a complex mix of serum proteins which are activated sequentially when antibodies in blood with immunity interact with the corresponding antigens. Complement proteins will quickly degrade if the plasma is not prepared and stored correctly. Our complement-preserved plasma is freshly collected aseptically and kept cold, and then flash-frozen at -80°C to prevent degradation of complement activity. We do not recommend this product be thawed for aliquoting, but we are able to accommodate custom volumes and aliquots upon request.
Catalog No. NC3588015 Supplier Innovative Research Inc Supplier No. ICPLAK2E50ML
Pooled Human Complement Plasma from Innovative Research is a complex mix of serum proteins which are activated sequentially when antibodies in blood with immunity interact with the corresponding antigens. Complement proteins will quickly degrade if the plasma is not prepared and stored correctly. Our complement-preserved plasma is freshly collected aseptically and kept cold, and then flash-frozen at -80°C to prevent degradation of complement activity. We do not recommend this product be thawed for aliquoting, but we are able to accommodate custom volumes and aliquots upon request.