Learn More
Pel Freez Biologicals Pooled Human OTC Serum (Neat) Heat Inactivated
Catalog No. NC3961258
Encompass
Human serum, pooled, 0.2 micron filtered, bottled, heat-inactivated for 30 minutes at 56°C and frozen.
Catalog No. NC3961258 Supplier Pel Freez Biologicals Supplier No. 340241
Human serum, pooled, 0.2 micron filtered, bottled, heat-inactivated for 30 minutes at 56°C and frozen.