Learn More
Innovative Research Inc Pooled Human Saliva 250ml
Catalog No. 502036058
Encompass_Preferred
Pooled Human Saliva from Innovative Research is collected fresh from two or more donors, pooled, and stored frozen. Produced in the salivary glands, human saliva is 99.5% water, but contains many important substances including electrolytes, mucus, antibacterial compounds, and various enzymes.
Catalog No. 50-203-6058 Supplier Innovative Research Inc Supplier No. IRHUSL250ML
Pooled Human Saliva from Innovative Research is collected fresh from two or more donors, pooled, and stored frozen. Produced in the salivary glands, human saliva is 99.5% water, but contains many important substances including electrolytes, mucus, antibacterial compounds, and various enzymes.