Learn More
Innovative Research Inc Pooled Human Saliva 50ml
Catalog No. 502036057
Encompass_Preferred
Pooled Human Saliva from Innovative Research is collected fresh from two or more donors, pooled, and stored frozen. Produced in the salivary glands, human saliva is 99.5% water, but contains many important substances including electrolytes, mucus, antibacterial compounds, and various enzymes.
Catalog No. 50-203-6057 Supplier Innovative Research Inc Supplier No. IRHUSL50ML
Pooled Human Saliva from Innovative Research is collected fresh from two or more donors, pooled, and stored frozen. Produced in the salivary glands, human saliva is 99.5% water, but contains many important substances including electrolytes, mucus, antibacterial compounds, and various enzymes.