Learn More
Innovative Research Inc Pooled Human Urine Filtered 1000ml
Supplier:  Innovative Research Inc IRHUUREFT1000ML
Encompass_Preferred
Pooled Human Urine Filtered from Innovative Research is 0.2 um filtered, pooled urine that has been prepared from a minimum of three unique donors. This product is suitable for use as a control for a wide range of in-vitro research applications.
Catalog No. 50-203-6014