Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Pooltester Reagent Refill Pack

Catalog No. 09050178 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-050-178 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-050-178 Supplier Lovibond™ Supplier No. 515980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains replacement reagents for the 5-in-1 Pool tester (PN 151900).

  • Includes 20x DPD No.1 Rapid tablets, 20x DPD No.3 Rapid tablets, 20x Phenol Red Rapid Tablets, 20x CyA-Test Tablets, 10k Alk Test Tablets, 10x Cal-Test tablets

Specifications

pH Range 6.8 to 8.2
Volume (Metric) 20 mL
Type Pooltester Reagents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.