Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSC Go Science Crazy Pop Beads
Shop All GSC Products
Click to view available options
Color:
Blue
Red
Yellow
Description
- Create DNA models
- Easily snaps into place
- Two-way
- Comes in pack of 150 beads
- Made from plastic
Specifications
Specifications
Description | Two-way beads |
Product Type | Pop Beads |
Quantity | 150 Beads/Pk. |
Material | Plastic |
Color | Red |
For Use With (Application) | Helps students learn and create models representing DNA strand, RNA conversion, nucleus |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction