Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Pop Beads

Catalog No. S28103 Shop All GSC Products
Change view
Click to view available options
Color:
Blue
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S28103 Yellow
S28101 Red
S28102 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options

Not a toy. Product intended for ages 13 and up.

Catalog No. S28103 Supplier GSC Go Science Crazy Supplier No. POPBDYL

Not a toy. Product intended for ages 13 and up.

For students to create cellular models related to DNA. Students learn about DNA and create models representing DNA strand, RNA conversion or nucleus.

  • Create DNA models
  • Easily snaps into place
  • Two-way
  • Comes in pack of 150 beads
  • Made from plastic

Specifications

Description Two-way beads
Product Type Pop Beads
Quantity 150 Beads/Pk.
Material Plastic
Color Yellow
For Use With (Application) Helps students learn and create models representing DNA strand, RNA conversion, nucleus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.