Learn More
Description
- Create DNA models
- Easily snaps into place
- Two-way
- Comes in pack of 150 beads
- Made from plastic
Specifications
Specifications
| Description | Two-way beads |
| Product Type | Pop Beads |
| Quantity | 150 Beads/Pk. |
| Material | Plastic |
| Color | Red |
| For Use With (Application) | Helps students learn and create models representing DNA strand, RNA conversion, nucleus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.