Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ POP™ Conformational Analysis Polymer (CAP)

Catalog No. 4340379
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-403-79 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-403-79 Supplier Applied Biosystems™ Supplier No. 4340379
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This POP™ polymer is a Conformational Analysis Polymer for non-denaturing polynucleotide fragment analysis applications on Genetic Analyzers.

This POP™ polymer is a Conformational Analysis Polymer for non-denaturing polynucleotide fragment analysis applications on Genetic Analyzers.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate at 2°C to 8°C.
For Use With (Equipment) 310 Genetic Analyzer, 3100 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3130 Genetic Analyzer, 3730 DNA Analyzer
For Use With (Application) Sequencing, Fragment Analysis (Non-Denaturing)
On Instrument Life Usage Stability on instrument 7 days
Product Type Sequencing Reagent
Quantity 25 mL
Shipping Condition Wet Ice
Do you offer any fragment analysis-based kits for non-denaturing applications?

We provide POP Conformational Analysis Polymer (CAP) for non-denaturing polynucleotide fragment analysis applications on the Applied Biosystems 3130/3130xl Genetic Analyzers. However, we do not have a premade kit specifically for this application. To learn more about non-denaturing applications, please see here (http://www.thermofisher.com/us/en/home/life-science/sequencing/fragment-analysis/single-strand-conformation-polymorphism-sscp.html).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.