Learn More
Research Products International Corp Pop-Off Cup for Vortex-Genie
SDP
Catalog No. 50998320
Encompass
Plastic pop-off cup for mixing and vortexing in single tubes. The pop-off cup may be used on the Vortex-Genie 1, Disruptor Genie, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers.
Catalog No. 50-998-320 Supplier Research Products International Corp Supplier No. 155311
Plastic pop-off cup for mixing and vortexing in single tubes. The pop-off cup may be used on the Vortex-Genie 1, Disruptor Genie, Vortex-Genie Pulse and the Vortex-Genie 2 family of mixers.