Learn More
OHAUS Pop-off Cup
Catalog No. 01804386 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-386 Vortex Attachment
1 options
Catalog No. 01-804-386 Supplier OHAUS Supplier No. 83041438
Add to Cart
Add to Cart
Pop-off cup for mixing and vortexing in single tubes.
  • The pop-off cup may be used on the Vortex Genie 1, Disruptor Genie, Vorex-Genie Pulse and the Vortex Genie 2 family of mixers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 1, Disruptor Genie, Vortex-Genie Pulse and the Vortex-Genie 2