Learn More
Electron Microscopy Sciences Pop-Up Slide Holder, 20 place, 1ea
SDP
Catalog No. 50931157
Encompass_Preferred
Holds 1x3" slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed.
Catalog No. 50-931-157 Supplier Electron Microscopy Sciences Supplier No. 71520
Holds 1x3" slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed.