Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pop-Up Tube Rack

Catalog No. 14127196 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Pink
Purple
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-196 Green
14-127-129 Purple
14-127-162 Blue
14-127-185 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-127-196
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Pop-Up Rack collapses for compact storage and pops up for quick use

  • Easy to open
  • Purple holds 15 mL tubes
  • Green holds 50 mL tubes
  • Blue and Pink hold both 15 and 50 mL tubes
  • Easy to close, stackable for compact storage
  • Alphanumeric well identification for quick location and retrieval of samples

Specifications

Autoclavable Not Autoclavable
Color Green
Holds Tubes (50 mL)
Material Polypropylene
Positions 1
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Product Type Test Tube Rack
Shelves 2 Tiers
Array 3 x 5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.