Learn More
AdipoGen POP1 human rec
SDP
Catalog No. 502562915
Encompass_Preferred
Protein. Human POP1 aa 2-89 is fused at the N-terminus to a GST tag. Source E. coli. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin IL-1beta and IL-18. Members of the Nod-like receptor family NLRs, including NLRP3/NALP3, are critical components of the inflammasome that link danger-signals to caspase-1 activation. Defects in NLRP3 are the cause of familial cold autoinflammatory syndrome type 1 FCAS1, Muckle-Wells syndrome MWS and of chronic infantile neurologic cutaneous and articular syndrome CINCA. Pyrin domain-containing protein 1 POP1 interacts with the protein Asc Pycard and modulates the interaction of Asc to NLR proteins.
Catalog No. 50-256-2915 Supplier AdipoGen Supplier No. AG40B0123C010
Protein. Human POP1 aa 2-89 is fused at the N-terminus to a GST tag. Source E. coli. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin IL-1beta and IL-18. Members of the Nod-like receptor family NLRs, including NLRP3/NALP3, are critical components of the inflammasome that link danger-signals to caspase-1 activation. Defects in NLRP3 are the cause of familial cold autoinflammatory syndrome type 1 FCAS1, Muckle-Wells syndrome MWS and of chronic infantile neurologic cutaneous and articular syndrome CINCA. Pyrin domain-containing protein 1 POP1 interacts with the protein Asc Pycard and modulates the interaction of Asc to NLR proteins.