Learn More
Aviva Systems Biology Corp POP5 Antibody - middle region (ARP56773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human POP5. Alternate names: HSPC004, RPP2, RPP20, hPop5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-209