Learn More
Medchemexpress LLC POPEth-d5 | 1612140-92-7 | 98.0% | 725.04 | 1 MG
SDP
Catalog No. 5000359872
Encompass_Preferred
POPEth-d5 is deuterium labeled POPEth. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-59872 Supplier Medchemexpress LLC Supplier No. HY146952S1MG
POPEth-d5 is deuterium labeled POPEth. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.