Learn More
Medchemexpress LLC POPEth-d5 | 1612140-92-7 | 98.0% | 725.04 | 500 UG
SDP
Catalog No. 5000359883
Encompass_Preferred
POPEth-d5 is deuterium labeled POPEth. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-59883 Supplier Medchemexpress LLC Supplier No. HY146952S500UG
POPEth-d5 is deuterium labeled POPEth. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.