Learn More
TARGETMOL CHEMICALS INC POPG sodium salt 25MG
SDP
Catalog No. 502880387
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. POPG sodium salt is a phospholipid with a negative charge and is a component of biological membranes. It can be used to study the function of biological membranes.
Catalog No. 50-288-0387 Supplier TARGETMOL CHEMICALS INC Supplier No. TF008525MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. POPG sodium salt is a phospholipid with a negative charge and is a component of biological membranes. It can be used to study the function of biological membranes.