Learn More
Medchemexpress LLC POPG (sodium salt) | 268550-95-4 | 99.2% | 50 MG
SDP
Catalog No. 5000339667
Encompass_Preferred
POPG sodium salt is a negatively charged phospholipid that influences the interactions of membrane proteins with other molecules by altering the charge characteristics of the lipid environment. It is useful in Parkinson's disease research and for creating nanoparticles for co-encapsulating and releasing hydrophilic peptide compounds.
  • Increases the apparent affinity of Gαq and Gβ1γ1 for activated NTS1
  • Can interact with positive charge of peptides
  • Bilayers formed can be penetrated by the antimicrobial peptide LL-37
  • Presence in the membrane enhances the binding of the PAP248-286 peptide
Catalog No. 50-003-39667 Supplier Medchemexpress LLC Supplier No. HY13046350MG
POPG sodium salt is a negatively charged phospholipid that influences the interactions of membrane proteins with other molecules by altering the charge characteristics of the lipid environment. It is useful in Parkinson's disease research and for creating nanoparticles for co-encapsulating and releasing hydrophilic peptide compounds.
  • Increases the apparent affinity of Gαq and Gβ1γ1 for activated NTS1
  • Can interact with positive charge of peptides
  • Bilayers formed can be penetrated by the antimicrobial peptide LL-37
  • Presence in the membrane enhances the binding of the PAP248-286 peptide