Learn More
Medchemexpress LLC POPG (sodium salt) | 268550-95-4 | 99.6% | C40H76NaO10P | 10MG
SDP
Catalog No. 5000259788
Encompass_Preferred
POPG (sodium salt) is a negatively charged phospholipid used to model anionic membrane environments and for liposome formation. It has molecular formula C40H76NaO10P, molecular weight 770.99, and CAS 268550-95-4, and is supplied as a solid suitable for laboratory membrane and biophysical studies.
  • Negatively charged phospholipid for modeling anionic membranes.
  • Suitable for liposome preparation and membrane reconstitution.
  • High purity (>99% HPLC) for reproducible results.
  • Molecular formula C40H76NaO10P, molecular weight 770.99.
  • Available as a solid for formulation and experimental use.
Catalog No. 50-002-59788 Supplier Medchemexpress LLC Supplier No. HY13046310MG
POPG (sodium salt) is a negatively charged phospholipid used to model anionic membrane environments and for liposome formation. It has molecular formula C40H76NaO10P, molecular weight 770.99, and CAS 268550-95-4, and is supplied as a solid suitable for laboratory membrane and biophysical studies.
  • Negatively charged phospholipid for modeling anionic membranes.
  • Suitable for liposome preparation and membrane reconstitution.
  • High purity (>99% HPLC) for reproducible results.
  • Molecular formula C40H76NaO10P, molecular weight 770.99.
  • Available as a solid for formulation and experimental use.