Learn More
Aviva Systems Biology Corp POR Antibody - N-terminal region (ARP44361_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44361_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human POR. Alternate names: CPR, CYPOR, DKFZp686G04235, FLJ26468, P450RApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58789