Learn More
Waters Corp Porapak N, Porous Polymer, 100 to 120 mesh, 35 g
Catalog No. 502391146 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Porapak N is a porous polymer designed for packed columns. It comes in a 100 to 120 mesh size and weighs 35 g.
  • Designed for packed columns
  • Suitable for the separation of amines
  • Suitable for the separation of hydrazines
  • Suitable for the separation of formaldehyde
  • Constructed from porous polymer
  • Features a 100 to 120 mesh size
  • Weighs 35 g
Catalog No. 50-239-1146 Supplier Waters Corp Supplier No. WAT027049
Waters Porapak N is a porous polymer designed for packed columns. It comes in a 100 to 120 mesh size and weighs 35 g.
  • Designed for packed columns
  • Suitable for the separation of amines
  • Suitable for the separation of hydrazines
  • Suitable for the separation of formaldehyde
  • Constructed from porous polymer
  • Features a 100 to 120 mesh size
  • Weighs 35 g