Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ QS, 50-80 mesh

Catalog No. 111004046 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4046 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4046 Supplier MilliporeSigma™ Supelco™ Supplier No. 20342
Add to Cart
Add to Cart

Porapak™™ QS treated with 2% phosphoric acid maybe used as column packing for gas chromatographic analysis of volatile fatty acids in rumen fluid.

Porapak™ porous polymers are spherical beads with consistent particle size, porosity, and surface area that help to insure analytical precision and accuracy.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 50/80 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜550 m2/g
Product Line PoraPak
For Use With (Application) GC
Density ˜0.34 g/mL (free fall density)

PoraPak is a trademark of Waters

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.