Learn More
MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ S, 100-120 mesh
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Particle Size | 100/120 Mesh |
| Technique | Chromatography |
| Packaging | Bottle |
| Quantity | 75 cc |
| Surface Area (Metric) | ˜375 m2/g |
| Product Line | PoraPak |
| Material | Crosslinked Polystyrene |
| For Use With (Application) | GC |
| Density | ˜0.35 g/mL (free fall density) |
PoraPak is a trademark of Waters
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.