Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ S, 80-100 mesh

Catalog No. 111004043 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4043 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4043 Supplier MilliporeSigma™ Supelco™ Supplier No. 20337
Add to Cart
Add to Cart

Porapak™™ S maybe used in isothermal gas chromatographic analysis of light gases, for detection and separation of CO2, C2H2, C2H4 and C2H6.

Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. Porapak™™ S is a porous polymer composed of vinyl pyridine and is most suitable for separation of normal and branched alcohols.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜375 m2/g
Product Line PoraPak
For Use With (Application) GC
Density ˜0.35 g/mL (free fall density)

PoraPak is a trademark of Waters

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.