Learn More
Waters Corp Waters Porapak Porous Packing, PS, 50-80 Mesh, 20 g
Catalog No. 50785281 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Porapak PS is a porous polymer bead packing material for gas chromatography, known for its distinct retention characteristics and being the second least polar Porapak type.
  • Offers partition properties of an extended liquid surface.
  • Provides sharp, symmetrical peaks and low retention volumes for water, alcohols, and glycols.
  • Ensures no bleed or adsorption of polar compounds.
  • High-temperature stability and consistent retention times.
  • Easy column packing with high efficiency and rapid overload recovery.
  • Allows adjustment of water elution time, preventing interference.
  • Composed of rigid, cross-linked polymers.
  • Max temperature: 250 °C.
Catalog No. 50-785-281 Supplier Waters Corp Supplier No. WAT027083
Porapak PS is a porous polymer bead packing material for gas chromatography, known for its distinct retention characteristics and being the second least polar Porapak type.
  • Offers partition properties of an extended liquid surface.
  • Provides sharp, symmetrical peaks and low retention volumes for water, alcohols, and glycols.
  • Ensures no bleed or adsorption of polar compounds.
  • High-temperature stability and consistent retention times.
  • Easy column packing with high efficiency and rapid overload recovery.
  • Allows adjustment of water elution time, preventing interference.
  • Composed of rigid, cross-linked polymers.
  • Max temperature: 250 °C.