Learn More
Waters Corp Waters Corporation GC Packing, Porapak PS, 80-100 Mesh, 20 g
Catalog No. 50785282 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a rigid, cross-linked polymer bead material designed for gas chromatography. It offers high-temperature stability, consistent retention times, and ensures no bleed from the packing material. This material helps achieve sharp, symmetrical peaks for a variety of compounds.
  • Absence of bleed from packing material
  • No adsorption of polar compounds
  • High-temperature stability
  • Consistent retention times
  • Ease of column packing with high efficiency
  • Rapid overload recovery
  • Rigid, cross-linked polymer beads
  • Provides sharp, symmetrical peaks and low retention volumes for water, alcohols, and glycols
  • Upper temperature limit is 250°C in an oxygen-free atmosphere
  • Particles do not crush or fracture easily during column packing
  • Columns can achieve plate counts of 1200 to 2200 plates per meter when properly packed
Catalog No. 50-785-282 Supplier Waters Corp Supplier No. WAT027084
This product is a rigid, cross-linked polymer bead material designed for gas chromatography. It offers high-temperature stability, consistent retention times, and ensures no bleed from the packing material. This material helps achieve sharp, symmetrical peaks for a variety of compounds.
  • Absence of bleed from packing material
  • No adsorption of polar compounds
  • High-temperature stability
  • Consistent retention times
  • Ease of column packing with high efficiency
  • Rapid overload recovery
  • Rigid, cross-linked polymer beads
  • Provides sharp, symmetrical peaks and low retention volumes for water, alcohols, and glycols
  • Upper temperature limit is 250°C in an oxygen-free atmosphere
  • Particles do not crush or fracture easily during column packing
  • Columns can achieve plate counts of 1200 to 2200 plates per meter when properly packed