Learn More
Waters Corp Porapak Bulk Media, QS, 50-80 Mesh, 26 g
Catalog No. 50785284 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This bulk media is a Porapak Porous Packing material of type QS, commonly used in chromatography applications.
  • Made of Porapak QS material.
  • Features a mesh size of 50 to 80.
  • Supplied in a 26 g quantity.
  • Suitable for chromatography applications.
Catalog No. 50-785-284 Supplier Waters Corp Supplier No. WAT027089
This bulk media is a Porapak Porous Packing material of type QS, commonly used in chromatography applications.
  • Made of Porapak QS material.
  • Features a mesh size of 50 to 80.
  • Supplied in a 26 g quantity.
  • Suitable for chromatography applications.