Learn More
Waters Corp Waters Porapak R Bulk Media, 80-100 Mesh, 24 g
Catalog No. 50785273 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Porapak R is a porous polymer resin designed for use as a stationary phase in gas chromatography. This bulk media facilitates the separation of various compounds, especially polar and volatile substances, making it suitable for a range of analytical applications.
  • Porous polymer resin for chromatographic separations
  • Optimized for gas chromatography applications
  • Effective for separating polar and volatile compounds
  • Specific particle size for consistent column packing
  • Bulk quantity provided for laboratory customization
Catalog No. 50-785-273 Supplier Waters Corp Supplier No. WAT027066
Porapak R is a porous polymer resin designed for use as a stationary phase in gas chromatography. This bulk media facilitates the separation of various compounds, especially polar and volatile substances, making it suitable for a range of analytical applications.
  • Porous polymer resin for chromatographic separations
  • Optimized for gas chromatography applications
  • Effective for separating polar and volatile compounds
  • Specific particle size for consistent column packing
  • Bulk quantity provided for laboratory customization