Learn More
Waters Corp Waters Corporation PoraPak Rxn CX 6 cc Vac Cartridge, 400 mg Sorbent, 80 μm
Catalog No. 50782475 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn CX is a polymer-based strong cation-exchange resin designed specifically for post synthesis cleanup of reaction mixtures in drug discovery.
  • Polymer-based strong cation-exchange resin for post-synthesis cleanup in drug discovery
  • CX chemistry with ion-exchange mode
  • Not endcapped and water wettable
  • Vac cartridge format with 6 cc barrel size
  • Ion exchange capacity of 1 meq/gram
  • Mass spec compatible
  • Particle size: 80 μm
  • Copolymer sorbent substrate
  • Sorbent weight: 400 mg
  • pH range: 0 to 14 pH
  • Available in 30 units per package
Catalog No. 50-782-475 Supplier Waters Corp Supplier No. 186004541
PoraPak Rxn CX is a polymer-based strong cation-exchange resin designed specifically for post synthesis cleanup of reaction mixtures in drug discovery.
  • Polymer-based strong cation-exchange resin for post-synthesis cleanup in drug discovery
  • CX chemistry with ion-exchange mode
  • Not endcapped and water wettable
  • Vac cartridge format with 6 cc barrel size
  • Ion exchange capacity of 1 meq/gram
  • Mass spec compatible
  • Particle size: 80 μm
  • Copolymer sorbent substrate
  • Sorbent weight: 400 mg
  • pH range: 0 to 14 pH
  • Available in 30 units per package