Learn More
Waters Corp Waters Vac Cartridge, 6 cc, Flangeless, 400 mg Sorbent, 80 μm
Catalog No. 50782476 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn CX is a polymer-based strong cation-exchange resin specifically designed for the post-synthesis cleanup of reaction mixtures in drug discovery.
  • Polymer-based strong cation-exchange resin
  • Cartridge volume: 6 cc
  • Flangeless vac cartridge format
  • Sorbent weight: 400 mg per cartridge
  • Particle size: 80 μm
  • 30 units per package
  • CX chemistry
  • Not endcapped
  • Ion exchange capacity: 1 meq/gram
  • Mass spec compatible
  • Ion-exchange mode
  • Copolymer sorbent substrate
  • Water wettable
  • pH range: 0 to 14 pH
Catalog No. 50-782-476 Supplier Waters Corp Supplier No. 186004542
PoraPak Rxn CX is a polymer-based strong cation-exchange resin specifically designed for the post-synthesis cleanup of reaction mixtures in drug discovery.
  • Polymer-based strong cation-exchange resin
  • Cartridge volume: 6 cc
  • Flangeless vac cartridge format
  • Sorbent weight: 400 mg per cartridge
  • Particle size: 80 μm
  • 30 units per package
  • CX chemistry
  • Not endcapped
  • Ion exchange capacity: 1 meq/gram
  • Mass spec compatible
  • Ion-exchange mode
  • Copolymer sorbent substrate
  • Water wettable
  • pH range: 0 to 14 pH