Learn More
Waters Corp Waters PoraPak Rxn CX Vac Cartridge, 60 cc, 5 g, 80 μm
Catalog No. 50782478 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn CX is a polymer-based strong cation-exchange resin for post-synthesis cleanup in drug discovery and concentrating high explosives. It substitutes for octadecyl-bonded silica and works with samples across a wide pH range.
  • Designed for post-synthesis cleanup in drug discovery.
  • Hydrophobic poly(divinylbenzene-vinylpyrrolidone) resin.
  • Concentrates high explosives in watery samples.
  • Substitutes for octadecyl-bonded silica.
  • Compatible with wide pH range (0 to 14).
  • High-caliber packings with reproducible activity and capacity.
  • Exceptional surface area, pore size, and particle-size distributions.
  • Facilitates easy extraction of samples.
  • Ideal for enriching and purifying samples for chromatography.
  • Non-contaminating poly-foil bag maintains purity.
Catalog No. 50-782-478 Supplier Waters Corp Supplier No. 186004544
PoraPak Rxn CX is a polymer-based strong cation-exchange resin for post-synthesis cleanup in drug discovery and concentrating high explosives. It substitutes for octadecyl-bonded silica and works with samples across a wide pH range.
  • Designed for post-synthesis cleanup in drug discovery.
  • Hydrophobic poly(divinylbenzene-vinylpyrrolidone) resin.
  • Concentrates high explosives in watery samples.
  • Substitutes for octadecyl-bonded silica.
  • Compatible with wide pH range (0 to 14).
  • High-caliber packings with reproducible activity and capacity.
  • Exceptional surface area, pore size, and particle-size distributions.
  • Facilitates easy extraction of samples.
  • Ideal for enriching and purifying samples for chromatography.
  • Non-contaminating poly-foil bag maintains purity.