Learn More
Waters Corp Waters Corporation Bulk Media, CX, 200 mL, 80 μm
Catalog No. 50782483 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn CX is a polymer-based strong cation-exchange resin specifically designed for the post-synthesis cleanup of reaction mixtures in drug discovery.
  • Polymer-based strong cation-exchange resin
  • Designed for post-synthesis cleanup of reaction mixtures in drug discovery
  • CX chemistry
  • 1 meq/gram ion exchange capacity
  • Ion-exchange separation mode
  • 80 μm particle size
  • Copolymer particle substrate
Catalog No. 50-782-483 Supplier Waters Corp Supplier No. 186004569
PoraPak Rxn CX is a polymer-based strong cation-exchange resin specifically designed for the post-synthesis cleanup of reaction mixtures in drug discovery.
  • Polymer-based strong cation-exchange resin
  • Designed for post-synthesis cleanup of reaction mixtures in drug discovery
  • CX chemistry
  • 1 meq/gram ion exchange capacity
  • Ion-exchange separation mode
  • 80 μm particle size
  • Copolymer particle substrate