Learn More
Waters Corp PoraPak Rxn Vac Cartridge, RP, 6 cc, 400 mg, 80 μm
Catalog No. 50782479 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn RP is a polymer-based reversed-phase resin designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Features reversed phase chemistry.
  • Not endcapped.
  • Vac cartridge format.
  • Mass spec compatible.
  • Copolymer sorbent substrate.
  • Sorbent weight of 400 mg.
  • Water wettable.
  • pH range of 0 pH to 14 pH.
  • Barrel size of 6 cc.
  • Particle size of 80 μm.
Catalog No. 50-782-479 Supplier Waters Corp Supplier No. 186004545
PoraPak Rxn RP is a polymer-based reversed-phase resin designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Features reversed phase chemistry.
  • Not endcapped.
  • Vac cartridge format.
  • Mass spec compatible.
  • Copolymer sorbent substrate.
  • Sorbent weight of 400 mg.
  • Water wettable.
  • pH range of 0 pH to 14 pH.
  • Barrel size of 6 cc.
  • Particle size of 80 μm.