Learn More
Waters Corp PoraPak Rxn RP Cartridge, 6 cc, Flangeless Vac, 400 mg Sorbent, 80 μm
Catalog No. 50782480 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn RP cartridges are polymer-based reversed-phase resins designed for the cleanup of synthetic reactions. They enable the separation of compounds based on polarity and are particularly useful for post-synthesis cleanup of neutral compounds from reaction mixtures in drug discovery. These cartridges offer increased speed, efficiency, and enhanced separation effectiveness compared to traditional cleanup methods.
  • Polymer-based reversed-phase resin for post-synthesis cleanup
  • Hard material resists increasing back pressure with flow
  • Minimal swelling or shrinking across solvents and pH extremes
  • Low hydraulic resistance for gravity flow
  • Tolerant to extreme pH without dissolution or hydrolysis
  • Robust, chemically resistant construction
  • Effective for fractionation and de-watering processes
  • Compatible with automated and manual setups
Catalog No. 50-782-480 Supplier Waters Corp Supplier No. 186004546
PoraPak Rxn RP cartridges are polymer-based reversed-phase resins designed for the cleanup of synthetic reactions. They enable the separation of compounds based on polarity and are particularly useful for post-synthesis cleanup of neutral compounds from reaction mixtures in drug discovery. These cartridges offer increased speed, efficiency, and enhanced separation effectiveness compared to traditional cleanup methods.
  • Polymer-based reversed-phase resin for post-synthesis cleanup
  • Hard material resists increasing back pressure with flow
  • Minimal swelling or shrinking across solvents and pH extremes
  • Low hydraulic resistance for gravity flow
  • Tolerant to extreme pH without dissolution or hydrolysis
  • Robust, chemically resistant construction
  • Effective for fractionation and de-watering processes
  • Compatible with automated and manual setups