Learn More
Waters Corp PoraPak Rxn RP 60 cc Vac Cartridge, 5 g Sorbent per Cartridge, 80 µm, 10/pk
Catalog No. 50782482 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn RP is a polymer-based reversed-phase resin designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
Catalog No. 50-782-482 Supplier Waters Corp Supplier No. 186004548
PoraPak Rxn RP is a polymer-based reversed-phase resin designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.