Learn More
Waters Corp WATERS CORPORATION PoraPak Rxn RP Bulk Media, 200 mL, 80 μm
Catalog No. 50782484 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PoraPak Rxn RP is a polymer-based reversed-phase resin designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Features a reversed phase chemistry.
  • Utilizes a copolymer particle substrate.
  • Particle size of 80 μm.
  • Provided as bulk media.
Catalog No. 50-782-484 Supplier Waters Corp Supplier No. 186004570
PoraPak Rxn RP is a polymer-based reversed-phase resin designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Designed for post synthesis cleanup of neutral compounds from reaction mixtures in drug discovery.
  • Features a reversed phase chemistry.
  • Utilizes a copolymer particle substrate.
  • Particle size of 80 μm.
  • Provided as bulk media.