Learn More
Waters Corp Waters Corporation Porapak Porous Packing, Type S, 50-80 Mesh
Catalog No. 50785275 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Porapak porous packings offer significant advantages in gas chromatography, featuring partition properties similar to a highly extended liquid surface. They eliminate common issues associated with support, polarity, liquid-phase volatility, or freezing point that often restrict gas/liquid chromatography. This design ensures sharp, symmetrical peaks and low retention volumes for substances including water, alcohols, and glycols.
  • No bleed from the packing material
  • No adsorption of polar compounds
  • High temperature stability
  • Consistent retention time
  • Easy column packing with high efficiency
  • Rapid overload recovery
  • Porous polymer beads modified for varied retention characteristics
  • Adjustable water peak position in chromatograms
Catalog No. 50-785-275 Supplier Waters Corp Supplier No. WAT027071
Waters Porapak porous packings offer significant advantages in gas chromatography, featuring partition properties similar to a highly extended liquid surface. They eliminate common issues associated with support, polarity, liquid-phase volatility, or freezing point that often restrict gas/liquid chromatography. This design ensures sharp, symmetrical peaks and low retention volumes for substances including water, alcohols, and glycols.
  • No bleed from the packing material
  • No adsorption of polar compounds
  • High temperature stability
  • Consistent retention time
  • Easy column packing with high efficiency
  • Rapid overload recovery
  • Porous polymer beads modified for varied retention characteristics
  • Adjustable water peak position in chromatograms