Learn More
Waters Corp Waters Porasil Radial-Pak PrepPAK Cartridge, Silica, 37 to 55 μm, 125 Å
Catalog No. 50785229 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This silica packing material provides long column life and low backpressure. Its high surface area makes it ideal for preparative work. The columns are designed as plastic compression cartridge segments for use in liquid chromatography systems with radial compression modules (RCM-holder sold separately).
  • Provides long column life and low backpressure
  • High surface area for preparative work
  • Designed as plastic compression cartridge segments for use in liquid chromatography systems
  • Normal-phase separation mode
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 bar)
  • Silica bonding technology
  • High silanol activity and irregular particle shape
  • Radial-Pak column format
  • HPLC system compatible
Catalog No. 50-785-229 Supplier Waters Corp Supplier No. WAT025853
This silica packing material provides long column life and low backpressure. Its high surface area makes it ideal for preparative work. The columns are designed as plastic compression cartridge segments for use in liquid chromatography systems with radial compression modules (RCM-holder sold separately).
  • Provides long column life and low backpressure
  • High surface area for preparative work
  • Designed as plastic compression cartridge segments for use in liquid chromatography systems
  • Normal-phase separation mode
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 bar)
  • Silica bonding technology
  • High silanol activity and irregular particle shape
  • Radial-Pak column format
  • HPLC system compatible