Learn More
Waters Corp Waters Corporation Porasil Silica Column, 125Å, 15-20μm, 3.9 mm X 300 mm
Catalog No. 50785245 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Porasil silica packing material offers long column life, low backpressure, and a high surface area. It is suitable for preparative work and normal-phase separations. Produced in a cGMP, ISO 9001-certified facility, the column uses ultra-pure reagents.
  • Offers long column life and low backpressure
  • High surface area suitable for preparative work
  • Silica-based material provides normal-phase separations for various sample types
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents
  • Chromatographically tested batches ensure repeatable performance
  • Intelligent chip provides performance chromatogram and certificate of batch analysis
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • 15 to 20 μm particle size
  • 125 Å pore size
  • 3.9 mm inner diameter
  • 300 mm length
Catalog No. 50-785-245 Supplier Waters Corp Supplier No. WAT025874
This Porasil silica packing material offers long column life, low backpressure, and a high surface area. It is suitable for preparative work and normal-phase separations. Produced in a cGMP, ISO 9001-certified facility, the column uses ultra-pure reagents.
  • Offers long column life and low backpressure
  • High surface area suitable for preparative work
  • Silica-based material provides normal-phase separations for various sample types
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents
  • Chromatographically tested batches ensure repeatable performance
  • Intelligent chip provides performance chromatogram and certificate of batch analysis
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • 15 to 20 μm particle size
  • 125 Å pore size
  • 3.9 mm inner diameter
  • 300 mm length