Learn More
Waters Corp Porasil Silica Radial-Pak Guard, 125Å, 15 - 20 µm, 40 mm X 10 mm, 2/pk
Catalog No. 50785439 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Porasil 15 - 20µm silica packing material provides long column life and low backpressure. The packing also has a high surface area making it ideal for preparative work.
Catalog No. 50-785-439 Supplier Waters Corp Supplier No. WAT037830
Waters Porasil 15 - 20µm silica packing material provides long column life and low backpressure. The packing also has a high surface area making it ideal for preparative work.