Learn More
Cole-Parmer Cole-Parmer Essentials Mortar and Pestle Set, Porcelain; 500 mL
Catalog No. NC1837106 Shop All Cole-Parmer Products
Encompass_Preferred
Mortars are glazed on the outside and pestles are glazed to grinding surface. Features pouring spout for easy pouring. Both mortar and pestle are sold as a set.
Catalog No. NC1837106 Supplier Cole-Parmer Supplier No. 6310060
Mortars are glazed on the outside and pestles are glazed to grinding surface. Features pouring spout for easy pouring. Both mortar and pestle are sold as a set.