Learn More
Raybiotech Inc Porcine CCL11 ELISA Kit
Supplier:  Raybiotech Inc ELPEOTAXIN1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.6 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Porcine CCL11.
Catalog No. 50-195-0182