Learn More
Raybiotech Inc C-Series Porcine Chemokine Array C1 Kit.
Supplier:  Raybiotech Inc AAPCHE18
Encompass_Preferred
Detects 12 Porcine Chemokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CCL3L1, Eotaxin-1, IFN alpha, IFN-beta, IFN-gamma, IL-1 alpha, IL-1 beta, MIF, MIG, MIP-1 beta, RANTES, TNF alpha
Catalog No. 50-194-8751