Learn More
Sino Biological Porcine circovirus 2 (PCV2) Capsid Antibody, Rabbit PAb, Antigen Affinity Purified 100 µL
Catalog No. 5000751263
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Porcine circovirus 2 (PCV2) Capsid (Catalog# 40552-V08B; Asn42-Pro233). Capsid specific IgG was purified by Porcine circovirus 2 (PCV2) Capsid affinity chromatography.
Catalog No. 50-007-51263 Supplier Sino Biological Supplier No. 40552T62100
Produced in rabbits immunized with purified, recombinant Porcine circovirus 2 (PCV2) Capsid (Catalog# 40552-V08B; Asn42-Pro233). Capsid specific IgG was purified by Porcine circovirus 2 (PCV2) Capsid affinity chromatography.