Learn More
Raybiotech Inc Porcine MIG (CXCL9) ELISA Kit
Supplier:  Raybiotech Inc ELPCXCL91
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This antibody pair shows no cross-reactivity with human, mouse, bovine and feline CXCL9/MIG
Catalog No. 50-195-0180